Corning™ Disposable Glass Pasteur Pipets
Feature a constriction one inch below the top to facilitate plugging. Corning™ Disposable Glass Pasteur Pipets are ideal for nonvolumetric transfer work in bacteriology, immunology, hematology and serology studies.
Manufacturer:  Corning™ 7095B9*
 View more versions of this product
Catalog No. 13-678-4A
Description
Description
Feature a constriction one inch below the top to facilitate plugging. Corning™ Disposable Glass Pasteur Pipets are ideal for nonvolumetric transfer work in bacteriology, immunology, hematology and serology studies.
  • Available in convenient bulk packs
  • Made from economical soda lime glass
  • One-time use eliminates the danger of cross-contamination of valuable specimens or laboratory reagents
  • For blood bank and clinical chemistry procedures
Specifications
Specifications
No
0.55cm
1 in. Constriction below the top
22.86cm
Unplugged Pasteur Pipet
0.216 in.
Yes
9 in.
Round
Soda Lime Glass
Documents