Corning™ Disposable Glass Pasteur Pipets
Feature a constriction one inch below the top to facilitate plugging. Corning™ Disposable Glass Pasteur Pipets are ideal for nonvolumetric transfer work in bacteriology, immunology, hematology and serology studies.
Manufacturer:  Corning™ C7095B5X
 View more versions of this product
Catalog No. 13-678-4
Description
Description
Feature a constriction one inch below the top to facilitate plugging. Corning™ Disposable Glass Pasteur Pipets are ideal for nonvolumetric transfer work in bacteriology, immunology, hematology and serology studies.
  • Available in convenient bulk packs
  • Made from economical soda lime glass
  • One-time use eliminates the danger of cross-contamination of valuable specimens or laboratory reagents
  • For blood bank and clinical chemistry procedures
Specifications
Specifications
No
0.65cm
1 in. Constriction below the top
14.6cm
Unplugged Pasteur Pipet
0.255 in.
Yes
5.75 in.
Round
Soda Lime Glass
Documents
Accessories