Please login to your online account to display your discounted pricing

Corning™ DeckWorks™ Tip Stations


Designed for single and multichannel pipetting stations

Manufacturer: corning™  C4144

 View more versions of this product

Catalog No. 10-320-733

  • / Pack of 960 Each
  • / Case of 5 Pack
 

Description & Specifications

Specifications

Product Type Tip Station
Volume (Metric) 1 to 200μL
Sterility Sterile
Color Natural
Graduated Yes
Shape Round
Material Polypropylene
Packaging Multipack

  • Contains 960 tips available in extended length 10μL and traditional 200μL sizes
  • Maximizes bench space while minimizing plastic waste
  • The 5-layer deck design (2 × 96) matches the microtiter plate format
  • This stable pipetting platform fits easily into sterile hood environments
  • Lids can be snap-positioned to expose a single side (96 tips) only
  • Applications include pre- and post-PCR procedures, cell culture and assays requiring large tip quantities
  • Either sterile or nonsterile