Please login to your online account to display your discounted pricing

Corning™ DeckWorks™ Reload Systems Starter Kits


Contains one DeckWorks Reload System with standard tips and empty hinged racks

Manufacturer: corning™  C4131

 View more versions of this product

Catalog No. 10-320-720

  • / Case of 960 Each
 

Description & Specifications

Specifications

Product Type Reload System Starter Kit
Volume (Metric) 1 to 200μL
Sterility Nonsterile
Color Natural
Graduated Yes
Shape Round
Material Polypropylene
Packaging Deck
For Use With (Application) Designed to allow to maximize storage space and help reduce the production and waste of plastic materials