Please login to your online account to display your discounted pricing
Corning™ DeckWorks™ Reload Systems with Standard Tips
Packaged in transparent trays for easy tip identification. Corning™ Deckworks™ Reload Systems with Standard Tips provide convenient and economical reloading of durable hinged racks.
Manufacturer: corning™  C4125
 View more versions of this product
Catalog No. 10-320-714
  • / Pack of 960 Each
  • / Case of 4 Pack
Description & Specifications
Specifications
Product Type Standard Tip
Volume (Metric) 0.1 to 10μL
Sterility Nonsterile
Color Natural
Graduated Yes
Autoclavable Yes
Tip Style Standard
Shape Round
Material Polypropylene
Packaging Deck: 960 Tips/Pack, 4 Packs/Case
  • Available in 10μL, 200μL, 300μL and 1,000μL sizes
  • Requires no transfer devices, saves space and minimize packaging waste
  • Pipet tips are made from virgin medical-grade resins
  • Reload decks and rack bases are manufactured from recycled polypropylene
  • Nonsterile decks and racks are steam autoclavable