Please login to your online account to display your discounted pricing

Corning™ DeckWorks™ Reload Systems with Low-Binding Tips


Allows convenient and economical reloading of durable Corning DeckWorks hinged racks

Manufacturer: corning™  C4154

 View more versions of this product

Catalog No. 10-320-741

  • / Case of 3840 Each
 

Description & Specifications

Specifications

Product Type Low Binding Tip
Material Polypropylene
Sterility Nonsterile
Graduated Yes
Shape Round
Packaging Deck
For Use With (Application) Allows convenient and economical reloading of durable Corning DeckWorks hinged racks
Volume (Metric) 1 to 200μL
Color Natural