Please login to your online account to display your discounted pricing

Corning™ DeckWorks™ Reload Systems with Low-Binding Tips


Allows convenient and economical reloading of durable Corning DeckWorks hinged racks

Manufacturer: corning™  C4155

 View more versions of this product

Catalog No. 10-320-742

  • / Case of 3840 Each
 

Description & Specifications

Specifications

Product Type Low Binding Tip
Material Polypropylene
Sterility Nonsterile
Shape Round
Packaging Deck
Graduated Yes
Volume (Metric) 1 to 300μL
Color Natural
For Use With (Application) Allows convenient and economical reloading of durable Corning DeckWorks hinged racks