Please login to your online account to display your discounted pricing

Corning™ DeckWorks™ Reload Systems with Low-Binding Tips


Allows convenient and economical reloading of durable Corning DeckWorks hinged racks

Manufacturer: corning™  C4153

 View more versions of this product

Catalog No. 10-320-740

  • / Pack of 960 Each
  • / Case of 4 Pack
 

Description & Specifications

Specifications

Product Type Low Binding Tip
Material Polypropylene
Sterility Nonsterile
Graduated Yes
Packaging Deck
Volume (Metric) 0.1 to 10μL
Shape Round
For Use With (Application) Allows convenient and economical reloading of durable Corning DeckWorks hinged racks
Color Natural

  • Available in 10μL, 200μL and 300μL sizes
  • Proprietary low binding technology reduces surface tension on the interior wall of the tip
  • Leaching and/or subsequent sample degradation associated with silicone-based tips are eliminated
  • Ideal for quantitative assays in which sample binding can significantly alter results
  • Hinged racks within this system are manufactured from recycled polypropylene
  • Specific applications include accurate and precise pipetting of expensive reagents, DNA, proteins and/or peptides