Please login to your online account to display your discounted pricing

Corning™ DeckWorks Pipet Tips


Universal tip fits in hinged DeckWorks racks that allows for easy rack stacking

Manufacturer: corning™  C4123

 View more versions of this product

Catalog No. 10-320-712

  • / Case of 3840 Each
 

Description & Specifications

Specifications

Sterility Sterile
Product Type Standard Tip
Color Natural
Packaging Hinged Rack
Shape Round
For Use With (Application) Universal tip fits in hinged DeckWorks racks that allows for easy rack stacking
Volume (Metric) 1 to 300μL