Please login to your online account to display your discounted pricing

Corning™ DeckWorks Pipet Tips


Universal tip fits in hinged DeckWorks racks that allows for easy rack stacking

Manufacturer: corning™  C4120

 View more versions of this product

Catalog No. 10-320-709

  • / Pack of 960 Each
  • / Case of 4 Pack
 

Description & Specifications

Specifications

Sterility Sterile
Product Type Standard Tip
Shape Round
For Use With (Application) Universal tip fits in hinged DeckWorks racks that allows for easy rack stacking
Tip Style Hinged Rack, Universal Fit
Volume (Metric) 0.1 to 10μL
Color Natural
Packaging Hinged Rack

  • Available in 10μL, 200μL, 300μL and 1000μL sizes
  • The 10μL, 300μL and 1000μL tips are extended length and allow improved access into micro (1.5mL) and standard conical bottom (15/50mL) centrifuge tubes
  • Robust 96-tip racks are designed for repeated steam autoclave cycles
  • Can be reused by removing the empty grey tip deck and replacing it with a new reload deck of 96 tips from the DeckWorks reload system
  • Allow maximize storage space and help reduce the production and waste of plastic materials
  • Hinged racks within this system are manufactured from recycled polypropylene
  • Corning racked 300μL DeckWorks are designed for popular 300μL multi-channel pipettors