Please login to your online account to display your discounted pricing

Corning™ DeckWorks Pipet Tips


Universal tip fits in hinged DeckWorks racks that allows for easy rack stacking

Manufacturer: corning™  C4116

 View more versions of this product

Catalog No. 10-320-705

  • / Case of 3840 Each
 

Description & Specifications

Specifications

Product Type Standard Tip
Sterility Nonsterile
Shape Round
Volume (Metric) 1 to 200μL
Color Natural
Packaging Hinged Rack
For Use With (Application) Universal tip fits in hinged DeckWorks racks that allows for easy rack stacking
Tip Style Hinged Rack, Universal Fit

  • Available in 10μL, 200μL, 300μL and 1000μL sizes
  • The 10μL, 300μL and 1000μL tips are extended length and allow improved access into micro (1.5mL) and standard conical bottom (15/50mL) centrifuge tubes
  • Robust 96-tip racks are designed for repeated steam autoclave cycles
  • Can be reused by removing the empty grey tip deck and replacing it with a new reload deck of 96 tips from the DeckWorks reload system
  • Allow maximize storage space and help reduce the production and waste of plastic materials
  • Hinged racks within this system are manufactured from recycled polypropylene
  • Corning racked 300μL DeckWorks are designed for popular 300μL multi-channel pipettors