Please login to your online account to display your discounted pricing

Corning™ DeckWorks™ Low-Binding Hinged Rack Pipet Tips


Designed for quantitative assays in which sample binding can significantly alter results

Manufacturer: corning™  C4147

 View more versions of this product

Catalog No. 10-320-734

  • / Case of 3840 Each
 

Description & Specifications

Specifications

Sterility Nonsterile
Material Polypropylene
Product Type Low Binding Tip
For Use With (Application) Designed for quantitative assays in which sample binding can significantly alter results
Packaging Hinged Rack
Volume (Metric) 0.1 to 10μL
Tip Style Hinged Rack
Shape Round
Color Natural
Graduated Yes