Please login to your online account to display your discounted pricing

Corning™ DeckWorks™ Low-Binding Hinged Rack Pipet Tips


Designed for quantitative assays in which sample binding can significantly alter results

Manufacturer: corning™  C4152

 View more versions of this product

Catalog No. 10-320-739

  • / Case of 3840 Each
 

Description & Specifications

Specifications

Material Polypropylene
Product Type Low Binding Tip
Sterility Sterile
For Use With (Application) Designed for quantitative assays in which sample binding can significantly alter results
Graduated Yes
Tip Style Hinged Rack
Shape Round
Volume (Metric) 1 to 300μL
Packaging Hinged Rack
Color Natural