Please login to your online account to display your discounted pricing

Corning™ DeckWorks™ Low-Binding Hinged Rack Pipet Tips


Designed for quantitative assays in which sample binding can significantly alter results

Manufacturer: corning™  C4151

 View more versions of this product

Catalog No. 10-320-738

  • / Case of 3840 Each
 

Description & Specifications

Specifications

Product Type Low Binding Tip
Material Polypropylene
Sterility Sterile
Graduated Yes
Tip Style Hinged Rack
Shape Round
Packaging Hinged Rack
For Use With (Application) Designed for quantitative assays in which sample binding can significantly alter results
Volume (Metric) 1 to 200μL
Color Natural