Please login to your online account to display your discounted pricing

Corning™ DeckWorks™ Low-Binding Hinged Rack Pipet Tips


Designed for quantitative assays in which sample binding can significantly alter results

Manufacturer: corning™  C4150

 View more versions of this product

Catalog No. 10-320-737

  • / Pack of 960 Each
  • / Case of 4 Pack
 

Description & Specifications

Specifications

Sterility Sterile
Product Type Low Binding Tip
Material Polypropylene
Shape Round
Tip Style Hinged Rack
Color Natural
For Use With (Application) Designed for quantitative assays in which sample binding can significantly alter results
Volume (Metric) 0.1 to 10μL
Packaging Hinged Rack
Graduated Yes