Please login to your online account to display your discounted pricing

Corning™ DeckWorks™ Low-Binding Hinged Rack Pipet Tips


Designed for quantitative assays in which sample binding can significantly alter results

Manufacturer: corning™  C4149

 View more versions of this product

Catalog No. 10-320-736

  • / Case of 3840 Each
 

Description & Specifications

Specifications

Material Polypropylene
Sterility Nonsterile
Product Type Low Binding Tip
Tip Style Hinged Rack
Color Natural
Packaging Hinged Rack
Graduated Yes
For Use With (Application) Designed for quantitative assays in which sample binding can significantly alter results
Shape Round
Volume (Metric) 1 to 300μL