Please login to your online account to display your discounted pricing

Corning™ DeckWorks™ Low-Binding Hinged Rack Pipet Tips, Nonsterile


Designed for quantitative assays in which sample binding can significantly alter results

Manufacturer: corning™  C4148

 View more versions of this product

Catalog No. 10-320-735

  • / Pack of 960 Each
  • / Case of 4 Pack
 

Description & Specifications

Specifications

Product Type Low Binding Tip
Volume (Metric) 1 to 200μL
Sterility Nonsterile
Color Natural
Graduated Yes
Tip Style Hinged Rack
Shape Round
Material Polypropylene
Packaging Hinged Rack
For Use With (Application) Designed for quantitative assays in which sample binding can significantly alter results