Please login to your online account to display your discounted pricing

Corning™ DeckWorks™ Low Binding Hinged Rack Barrier Tips


Features an inert, high-density polyethylene barrier to eliminate aerosol carryover contamination during critical pipetting procedures

Manufacturer: corning™  C4137

 View more versions of this product

Catalog No. 10-320-726

  • / Pack of 960 Each
  • / Case of 4 Pack
 

Description & Specifications

Specifications

Quantity 960 tips/Pk.; 4 Pack/Cs.
Sterility Sterile
Product Type Low Binding Barrier Tip
Material HDPE
Packaging Hinged Rack
For Use With (Application) Used for quantitative assays in which sample binding can significantly alter results
Graduated Yes
Tip Style Barrier, Hinged Rack
Shape Round
Volume (Metric) 1 to 100μL
Color Natural