Please login to your online account to display your discounted pricing

Corning™ DeckWorks™ Low Binding Hinged Rack Barrier Tips


Features an inert, high-density polyethylene barrier to eliminate aerosol carryover contamination during critical pipetting procedures

Manufacturer: corning™  C4136

 View more versions of this product

Catalog No. 10-320-725

  • / Pack of 960 Each
  • / Case of 4 Pack
 

Description & Specifications

Specifications

Product Type Low Binding Barrier Tip
Volume (Metric) 1 to 20μL
Sterility Sterile
Color Natural
Graduated Yes
Tip Style Barrier, Hinged Rack
Shape Round
Material HDPE
Packaging Hinged Rack
Quantity 960 Tips/Pack, 4 Packs/Case
For Use With (Application) Used for quantitative assays in which sample binding can significantly alter results

  • The hinged rack design allows easy rack stacking
  • Racks can be reused
  • Simply remove the tip insert in the rack and replace with the Corning tip inserts for hinged racks
  • Proprietary low binding technology reduces surface tension on the interior wall of the tip
  • Allow maximize storage space and help reduce the production and waste of plastic materials
  • Hinged racks within this system are manufactured from recycled polypropylene
  • Used for quantitative assays in which sample binding can significantly alter results
  • These sterile tips will not trap liquids or inhibit PCR in the event of over-pipetting

Model No. 10320728: The unique design of this extended-length tip allows standardization across wide range of 20mL, 50mL, 100mL and 200mL pipettors.