Please login to your online account to display your discounted pricing

Corning™ DeckWorks™ Low Binding Hinged Rack Barrier Tips


Features an inert, high-density polyethylene barrier to eliminate aerosol carryover contamination during critical pipetting procedures

Manufacturer: corning™  C4135

 View more versions of this product

Catalog No. 10-320-724

  • / Pack of 960 Each
  • / Case of 4 Pack
 

Description & Specifications

Specifications

Sterility Sterile
Quantity 960 tips/Pk.; 4 Pack/Cs.
Product Type Low Binding Barrier Tip
Material HDPE
Volume (Metric) 0.1 to 10μL
Graduated Yes
Packaging Hinged Rack
Shape Round
Tip Style Barrier, Hinged Rack
Color Natural
For Use With (Application) Used for quantitative assays in which sample binding can significantly alter results

  • The hinged rack design allows easy rack stacking
  • Racks can be reused
  • Simply remove the tip insert in the rack and replace with the Corning tip inserts for hinged racks
  • Proprietary low binding technology reduces surface tension on the interior wall of the tip
  • Allow maximize storage space and help reduce the production and waste of plastic materials
  • Hinged racks within this system are manufactured from recycled polypropylene
  • Used for quantitative assays in which sample binding can significantly alter results
  • These sterile tips will not trap liquids or inhibit PCR in the event of over-pipetting

Model No. 10320728: The unique design of this extended-length tip allows standardization across wide range of 20mL, 50mL, 100mL and 200mL pipettors.