Please login to your online account to display your discounted pricing

Corning™ DeckWorks™ Bulk Pipet Tips


Compatible with DeckWorks racks and packaged in resealable polyethylene bags

Manufacturer: corning™  C4110

 View more versions of this product

Catalog No. 10-320-700

  • / Pack of 1000 Each
  • / Case of 10 Pack
 

Description & Specifications

Specifications

Product Type Tip
Material Polypropylene
For Use With (Equipment) DeckWorks racks
Sterility Nonsterile
Tip Style Disposable, Round
For Use With (Application) Help reduce the production and waste of plastic materials
Graduated Yes
Shape Round
Packaging Bulk
Volume (Metric) 0.1 to 10μL
Color Natural