missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Corning™ DeckWorks™ Bulk Pipet Tips
Compatible with DeckWorks racks and packaged in resealable polyethylene bags
$340.36 - $3,345.64
Specifications
For Use With (Equipment) | DeckWorks racks |
---|---|
Material | PP (Polypropylene) |
Packaging | Bulk |
Sterility | Non-sterile |
For Use With (Application) | Help reduce the production and waste of plastic materials |
Catalog Number | Mfr. No. | Volume (Metric) | Color | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Color | Price | Quantity | ||||
10-320-702
|
Corning™
C4112 |
1 to 200 μL | Yellow |
Case of 10000 Each for $3,345.64
|
|
10-320-700
|
Corning™
C4110 |
0.1 to 10 μL | Natural |
Pack of 1000 Each for $340.36
Case of 10 Pack for $3,403.60 |
|
10-320-703
|
Corning™
4114 |
100 to 2000 μL | Natural |
Pack of 768 Each for $347.91
Case of 10 Pack for $3,479.10 |
|
10-320-701
|
Corning™
C4111 |
1 to 200 μL | Natural |
Case of 10000 Each for $3,345.64
|
|
Description
Corning DeckWorks bulk pipet tips are compatible with DeckWorks racks and packaged in resalable polyethylene bags. Tip sizes include 10 μL, 200 μL, and 1000 μL and offer maximum savings.
Specifications
DeckWorks racks | |
Bulk | |
Help reduce the production and waste of plastic materials | |
Round |
PP (Polypropylene) | |
Non-sterile | |
Yes | |
Yes |