Corning™ DeckWorks™ Bulk Pipet Tips
Compatible with DeckWorks racks and packaged in resealable polyethylene bags
Manufacturer: Corning™ C4111
Description
Corning DeckWorks bulk pipet tips are compatible with DeckWorks racks and packaged in resalable polyethylene bags. Tip sizes include 10 μL, 200 μL, and 1000 μL and offer maximum savings.
Specifications
Yes | |
Yes | |
Round | |
Non-sterile | |
Help reduce the production and waste of plastic materials |
DeckWorks racks | |
PP (Polypropylene) | |
1 to 200 μL | |
Natural | |
Bulk |