Please login to your online account to display your discounted pricing
Corning™ Untreated Culture Dishes
Binds bio-molecules through passive interactions and hydrophobic in nature. Corning™ Untreated Culture Dishes provide an ideal environment for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface.
Manufacturer: corning™  C430597
 View more versions of this product
Catalog No. 08-772-25
  • / Case of 60 Each
Description & Specifications
Specifications
Surface Area (Metric) 152cm2
Height (Metric) Exterior 25mm
Material Optically Clear Virgin Polystyrene (PS)
Diameter (English) 5.9 in.
Diameter (Metric) 150mm
Sterile Yes
  • Manufactured from optically-clear virgin polystyrene
  • Not treated for cell attachement
  • Sterilized by gamma radiation and certified nonpyrogenic
  • Stacking beads to aid in handling
  • Supplied with vents to provide consistent gas exchange