Corning™ Cryogenics Workstation
Rack grips Corning externally and internally threaded self-standing vials, so caps can be removed with one hand
$194.41 - $252.72
Specifications
Color White
Holds 30 cryogenic vial
Material Polycarbonate
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
03-374-37B Corning™
C430526
Cryogenic Vial Rack Case for $194.41
03-374-37A Corning™
C430525
Cryogenic Workstation Case for $252.72
Specifications
Description
  • Reusable polycarbonate rack and storage tray
  • Removable rack has an alphanumeric grid and holds 30 vials
  • Tray can also be used to hold water or crushed ice
  • Rack and tray can be autoclaved at 121°C (250°F)
Order Info
Workstation dimensions: 14.63L × 5.69W × 3 in. H (37.1 × 14.5 × 7.6cm)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
White
30 cryogenic vial
Polycarbonate