Corning™ Costar™ Transtar™-96 96-Well Liquid Transfer System Accessories: Reservoir Liners
For use with 96-well plates
Manufacturer:  Corning™ C4878
 View more versions of this product
Catalog No. 07-200-58
Description
Description
For use with 96-well plates
These are sterile replacement open reservoir liners for the Transtar-96™ Well Liquid Transfer system.
Specifications
Specifications
8
1/Pk.
White
Polystyrene
96
100/Cs.
8 Channel Reservoir
Ideal for repetitively filling multi-channel pipettors
Sterile