Please login to your online account to display your discounted pricing

Corning™ Collagen IV, Mouse


Used as either a thin layer on tissue-culture surfaces to enhance cell attachment and proliferation, or as a gel to promote expression of cell-specific morphology and function

Manufacturer: corning™  C356233

 View more versions of this product

Catalog No. CB-40233B

  • / Each
 

Description & Specifications

Specifications

Formulation Frozen in 100mM borate buffer pH 8.0
Quantity 10mg
Type Collagen IV, Mouse
pH 8
Concentration 1 to 5μg/cm2
For Use With (Application) Used as a thin coating on tissue-culture surfaces to promote cell attachment and proliferation and to study the collagen's effects on cell behavior
Storage Requirements -20°C
Source Engelbreth-Holm-Swarm lathrytic Mouse Tumor