Please login to your online account to display your discounted pricing

Corning™ Collagen IV, Human


Used as either a thin layer on tissue-culture surfaces to enhance cell attachment and proliferation, or as a gel to promote expression of cell-specific morphology and function

Manufacturer: corning™  C354261

 View more versions of this product

Catalog No. 08-774-390

  • / Each
 

Description & Specifications

Specifications

pH 8
Quantity 500μg
Source Human Placenta
Formulation Sodium chloride 1M, Tris 1.25mM, pH 8
Concentration 1 to 5μg/cm2
For Use With (Application) Used as either a thin layer on tissue-culture surfaces to enhance cell attachment and proliferation, or as a gel to promote expression of cell-specific morphology and function
Storage Requirements -20°C
Type Collagen VI, Human