missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ Collagen IV, Human

Used as either a thin layer on tissue-culture surfaces to enhance cell attachment and proliferation, or as a gel to promote expression of cell-specific morphology and function

$1,151.80 - $1,477.39

Specifications

Concentration 1 to 5μg/cm2
Source Human Placenta
pH 8
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Formulation Storage Requirements Price Quantity  
Catalog Number Mfr. No. Quantity Formulation Storage Requirements Price Quantity  
08-774-390 View Documents Corning™
C354261
500μg Sodium chloride 1M, Tris 1.25mM, pH 8 -20°C Each for $1,477.39
 
CB-40245 View Documents Corning™
C354245
0.25mg Frozen in 100mm borate buffer pH 8.0 -70°C Each for $1,151.80
 
Description

Description

  • Source: Human placenta
  • Source material tested for HBsAG, for anti-HIV-1 and anti-HIV-2, and anti-HCV virus-1 and virus-2
  • Electrophoretically homogeneous (SDS-PAGE)
  • Formulation: Frozen in 100mm borate buffer pH 8.0

Specifications

Specifications

1 to 5μg/cm2
Human Placenta
8