Please login to your online account to display your discounted pricing

Corning™ Collagen I, Rat


Enhances cell attachment and proliferation or promotes expression of cell-specific morphology and function. Corning™ Collagen I, Rat is effective as a gel or a thin coating at a recommended concentration of 5 to 10μg/cm2 of growth surface depending on cell type.

Manufacturer: corning™  C356236

 View more versions of this product

Catalog No. CB-40236B

  • / Each
 

Description & Specifications

Specifications

Purity >90% by SDS-PAGE
For Use With (Application) Used either as a thin layer on tissue-culture surfaces to enhance cell attachment and proliferation, or as a gel to promote expression of cell-specific morphology and function
Concentration 3 to 4mg/mL
Type Collagen I, Rat Tail
Source Rat Tail Tendon
Quantity 1000mg
Formulation Liquid in 0.02N acetic acid
Storage Requirements 2° to 8°C

 

  • Source: Rat tail tendon
  • Purity: >90% by SDS-PAGE
  • Formulation: Liquid in 0.02 N acetic acid, concentration range 3 to 4mg/mL
  • Tested for: ability to promote attachment and spreading of HT-1080 human fibrosarcoma cells
  • Forms firm gel up to a dilution of 1:10; further dilution will decrease rigidity of gel
  • Tested and found negative for bacteria, fungi, and mycoplasma