missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ Collagen I, Human

Used as either a thin layer on tissue-culture surfaces to enhance cell attachment and proliferation, or as a gel to promote expression of cell-specific morphology and function

Manufacturer:  Corning™ C354243

 View more versions of this product

Catalog No. CB-40243


Alert:

In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.


Description

Description

  • Shown to promote attachment and spreading of HT-1080 human fibrosarcoma cells
  • Source: Human placenta
  • Electrophoretically homogeneous (SDS-PAGE)
  • Formulation: Frozen in 2mM Hcl

Specifications

Specifications

Collagen I, Human
Used as either a thin layer on tissue-culture surfaces to enhance cell attachment and proliferation, or as a gel to promote expression of cell-specific morphology and function
>90% by SDS-PAGE
0.25mg
0.2 to 2μg/cm2
Frozen in 2mM hydrogen chloride
-20°C
Human Placenta
Documents