Please login to your online account to display your discounted pricing

Corning™ Collagen I, Human


Used as either a thin layer on tissue-culture surfaces to enhance cell attachment and proliferation, or as a gel to promote expression of cell-specific morphology and function

Manufacturer: corning™  C354265

 View more versions of this product

 Alert

In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.

Catalog No. 08-774-550

  • / Each
 

Description & Specifications

Specifications

Concentration 0.2 to 2μg/cm2
For Use With (Application) Used as either a thin layer on tissue-culture surfaces to enhance cell attachment and proliferation, or as a gel to promote expression of cell-specific morphology and function
Formulation Frozen in 2mM hydrogen chloride
Storage Requirements -20°C
Quantity 10mg
Source Human Placenta
Type Collagen I, Human