missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Corning™ Clear Polypropylene 96-Well Expanded-Volume Plates
Offer reduced space requirements while maintaining full instrument compatibility
Manufacturer: Corning™ C3344
Description
Corning polypropylene microplates offer both small volume and large volume (blocks) well formats to meet assay and storage requirements.
- Flat, round, or V-shaped well bottom
- Features uniform skirt heights for greater robotic gripping surface
- Solvent resistant polypropylene provides compatibility with many common organic solvents (e.g., DMSO, ethanol, methanol)
- RNase-/DNase-free
- Available sterile or nonsterile
Order Info
Packaged in bags of ten
Certifications
Certified DNase- and RNase-free
Specifications
Standard | |
5 inner packs of 10 | |
450 μL | |
V | |
Clear | |
50/Cs. | |
10/Pk. | |
Non-treated | |
8 x 12 |
Without Lid | |
96-well Assay Microplate | |
V-bottom | |
DNase, RNase-free | |
Polypropylene | |
50/Cs. | |
Sterile | |
96 |