Corning™ Class A Reusable PMP Volumetric Flasks
Autoclaved to 121°C.
$2,475.80 - $2,560.65
Specifications
Material Perfluoroalkoxy Copolymer
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
S02289C Corning™
5650P100EMD
100mL Case of 6 Each for $2,560.65
S02289E Corning™
5650P500EMD
500mL Case of 4 Each for $2,475.80
Specifications
Description
  • Perfluoroalkoxy screw cap
  • Screw caps provide hermetic seals, reducing the possibility of outside contamination
  • Withstand high temperatures and are resistant to a variety of aqueous solutions and organic solvents
  • Translucent flask
  • Height dimension is for flask only
Description & Specifications
Specifications
Perfluoroalkoxy Copolymer