PYREX™ Chromatography Column With Coarse Fritted Disc
Designed for general chromatographic analysis
Manufacturer: PYREX™ C3845040
Description
These tubes are designed for general chromatographic analysis. The tops are tooled for rubber stopper fit and the tubes are fabricated with sealed-in coarse porosity fritted discs. The outflow tubes are large in diameter to eliminate the smearing of the definition bands of effluent caused by the tube being filled, and the solvent mixed before being expelled. The outflow tubes are long enough to protrude through a rubber stopper.
Specifications
General chromatographic analysis | |
Chromatography Column | |
750 mL | |
Borosilicate Glass | |
10 |
44 mm | |
40 to 60 μm | |
600 mm | |
Fritted disc |