Please login to your online account to display your discounted pricing

Corning™ CELLSTACK™ Culture Chambers


Featuring USP Class VI polystyrene construction for excellent optical clarity and mechanical strength. Corning™ CELLSTACK™ are available in five sizes and with two unique surface treatments in addition to the standard tissue culture treated surface.

Manufacturer: corning™  C3303

 View more versions of this product

Catalog No. 08-757-226

  • / Case of 8 Each
 

Description & Specifications

Specifications

Closure Color Orange
Diameter (English) Cap 1.29 in.
Diameter (Metric) Cap 33mm
Graduated Yes
Product Type Culture Chamber
Surface Treatment Ultra-Low Attachment
Volume (English) 4.22 to 6.42 oz.
Volume (Metric) 125 to 190mL
Culture Area 636cm2
Material Polystyrene
Sterilization Method Gamma Irradiated
Certifications/Compliance USP Class VI
Quantity One chamber
Sterile Yes

  • Two generous 26mm filling ports provide direct access to chamber bottom and flexibility for sterile filling and emptying
  • Standard 33mm threaded caps have 0.2μm pore nonwettable membranes sealed to caps—allowing gas exchange, preventing pressure buildup, and minimizing risk of contamination
  • Choice of surface treatments: standard Tissue Culture Treated Surface, CellBIND Surface and Ultra-Low Attachment Surface
  • Sterilized by gamma irradiation
  • Leak tested

  • Corning CellBIND™ surface treatment improves cell attachment leading to increased cell growth and yields and provides more consistent and even cell attachment. Surface helps adapt cells to reduced serum or serum-free conditions, requires no refrigeration or special handling and is stable at room temperature.
  • Ultra-Low attachment surface features a covalently bound hydrogel layer that effectively inhibits cellular attachment and minimizes protein absorption, enzyme activation and cellular activation