Please login to your online account to display your discounted pricing
Corning™ cellgro™ X-GAL and IPTG, Powder
Produced under highest industry standards
$74.20 - $459.78
Specifications
Quantity 1000mg
View More Specs
Products
Catalog Number Mfr. No. For Use With (Application) Price Quantity    
MT46101RF
corning™
MT46101RF
Research use only Each for $459.78
MT46102RF
corning™
46102RF
For screening clones when used with X-Gal to detect lac gene activity Each for $74.20
Description & Specifications
Specifications
Quantity 1000mg
X-Gal (5-bromo-4-chloro-3-indolyl-b-D-galactopyranoside)
  • Substrate of b-galactosidase
  • Hydrolyzed by β-galactosidase to produce indoxyl in bacterial medium
IPTG (isopropylthiogalactoside)
  • Inducer of lac operon in bacteria
  • Used with X-gal for detecting lac gene activity
  • For screening of clones containing gene of interest in lac operon