Please login to your online account to display your discounted pricing

Corning™ cellgro™ Cell Culture Grade Water Tested to USP Sterile Water


High quality water for use in cell culture applications. Corning™ cellgro™ Cell Culture Grade Water Tested to USP Sterile Water are used in preparation of purification and chromatography buffers.

Manufacturer: corning™  25055LH

 View more versions of this product

 Alert

Manufactured under cGMP (21 CFR 820) in ISO 13485 certified facility

Catalog No. MT25055LH

  • / Each
 

Description & Specifications

Specifications

pH 5 to 7
Sterility Yes
Quantity 1 x 200L
Endotoxin ≤0.025EU/mL
Purity Grade WFI Grade
Recommended Storage 2° to 30°C
Additional Information Used in cell culture, hydration of media, supplements, and salt solutions, device processing, process validation, preparation of purification and chromatography buffers
Storage Requirements 2-30°C
Product Type Cell Culture Grade Water
For Use With (Application) Used in cell culture, hydration of media, supplements, and salt solutions, device processing, process validation, preparation of purification and chromatography buffers
Endotoxin Concentration ≤0.025EU/mL
pH 5 to 7
Sterile Yes
Sterile Yes
Quantity 1 x 200L

  • Tested to USP monograph for sterile water for injection (WFI)
  • Ideal for the preparation of reagents, rinsing glassware and plasticware, and other molecular biology applications where RNase, DNase, and Protease-free water is requiredFree of any added components
  • No toxic agents, including DEPC, are used, eliminating possible interferences with enzymatic reactions
  • Endotoxin levels ≤0.025EU/mL
  • Available in product sizes ranging from 100mL to 200L

Accessories

2° to 30°C