Please login to your online account to display your discounted pricing

Corning™ cellgro™ Cell Culture Grade Water Tested to USP Sterile Water


High quality water for use in cell culture applications. Corning™ cellgro™ Cell Culture Grade Water Tested to USP Sterile Water are used in preparation of purification and chromatography buffers.

Manufacturer: corning™  25055CI

 View more versions of this product

 Alert

Manufactured under cGMP (21 CFR 820) in ISO 13485 certified facility

Catalog No. MT25055CI

  • / Pack of 6 Each
 

Description & Specifications

Specifications

Storage Requirements 2-30°C
Product Type Cell Culture Grade Water
Endotoxin Concentration ≤0.025EU/mL
For Use With (Application) Used in cell culture, hydration of media, supplements, and salt solutions, device processing, process validation, preparation of purification and chromatography buffers
pH 5 to 7
Quantity 6 x 100mL
Sterile Yes
Sterile Yes
Quantity 6 x 100mL

  • Tested to USP monograph for sterile water for injection (WFI)
  • Ideal for the preparation of reagents, rinsing glassware and plasticware, and other molecular biology applications where RNase, DNase, and Protease-free water is requiredFree of any added components
  • No toxic agents, including DEPC, are used, eliminating possible interferences with enzymatic reactions
  • Endotoxin levels ≤0.025EU/mL
  • Available in product sizes ranging from 100mL to 200L