Please login to your online account to display your discounted pricing

Corning™ cellgro™ Trace Elements


For Research applications

Manufacturer: corning™  25-022-CI

 View more versions of this product

Catalog No. MT99175CI

  • / Each
 

Description & Specifications

Specifications

Solution Type Trace Element B, 1000x
Quantity 100mL
Sterilization Method Sterile-filtered
Storage Requirements 2° to 8°C
Packaging Plastic Bottle
Format Liquid

cellgro incorporates many principles developed by the pharmaceutical industry including the use of WFI quality water, raw material testing, in-process monitoring, and finished product testing.

  • Trace Elements A includes Cupric Sulfate, Ferric Citrate, Sodium Selenite, and Zinc Sulfate
  • Trace Elements B includes Ammonium Molybdate, Ammonium Vanadate, Manganese Sulfate, Nickel Sulfate, Sodium Silicate, Stannous Chloride and Hydrochloric Acid
  • Trace Elements C includes Aluminum Chloride, Barium Acetate, Cadmium Chloride, Chromic Chloride, Cobalt Dichloride, Germanium Dioxide, Potassium Bromide, Potassium Iodide, Rubidium Chloride, Silver Nitrate, Sodium Fluoride, and Zirconyl Chloride