Please login to your online account to display your discounted pricing

Corning™ cellgro™ Penicillin-Streptomycin Solution


Effective against Gram-positive and Gram-negative bacteria. Corning™ cellgro™ Penicillin-Streptomycin Solution is a mix of the antibiotics Penicillin (10,000 IU) and Streptomycin (10,000µg/mL) in a 100-fold working concentration.

Manufacturer: corning™  30001CI

 View more versions of this product

Catalog No. MT30001CI

  • / Pack of 6 Each
 

Description & Specifications

Specifications

Storage -20°C
Packaging Plastic Bottle
Storage Requirements -20°C
Format Solutiion
Solution Type Penicillin-Streptomycin Solution, 50X
Sterile Yes
Concentration 5000 IU Penicillin, 5000μg/mL Streptomycin
Quantity 6 x 100mL

  • Certificate of Analysis available
  • Penicillin (Penicillin G) works by inhibiting peptidoglycan synthesis, while Streptomycin inhibits protein synthesis