Please login to your online account to display your discounted pricing

Corning™ cellgro™ Penicillin-Streptomycin Solution


Effective against Gram-positive and Gram-negative bacteria. Corning™ cellgro™ Penicillin-Streptomycin Solution is a mix of the antibiotics Penicillin (10,000 IU) and Streptomycin (10,000µg/mL) in a 100-fold working concentration.

Manufacturer: corning™  30002CI

 View more versions of this product

Catalog No. MT30002CI

  • / Pack of 6 Each
 

Description & Specifications

Specifications

Storage -20°C
Sterile Yes
Solution Type Penicillin-Streptomycin Solution, 100X
Concentration 10,000 IU Penicillin, 10,000μg/mL Streptomycin
Storage Requirements -20°C
Quantity 6 x 100mL
Sterile Yes
Packaging Type Plastic Bottle
Format Solutiion
Packaging Plastic Bottle
Quantity 6 x 100mL
Format Solutiion
Concentration 10,000 IU Penicillin, 10,000μg/mL Streptomycin

  • Certificate of Analysis available
  • Penicillin (Penicillin G) works by inhibiting peptidoglycan synthesis, while Streptomycin inhibits protein synthesis