Please login to your online account to display your discounted pricing

Corning™ cellgro™ Microbiological Media


Sterile filtered

Manufacturer: corning™  46050CM

 View more versions of this product

Catalog No. MT46050CM

  • / Pack of 6 Each
 

Description & Specifications

Specifications

Packaging Bottle
Fill Volume 6 x 1000mL
Manufacturing Origin United States
Media Type LB Broth
Quantity 6 x 1000mL
Format Liquid
For Use With (Application) For maintenance/propagation of Escherichia coli with plasmid DNA and recombinant Protein production
Media Format Casein Peptone 10g/L, NaCl 10g/L