Please login to your online account to display your discounted pricing

Corning™ cellgro™ Lymphocyte Separation Medium


Designed for the in vitro isolation of lymphocytes from diluted whole blood

Manufacturer: corning™  25072CI

 View more versions of this product

Catalog No. MT25072CI

  • / Each
 

Description & Specifications

Specifications

Sterile Yes
Quantity 1 x 100mL
Storage Requirements 15° to 30°C
Format Liquid
Product Type Lymphocyte separation medium