Please login to your online account to display your discounted pricing

Corning™ cellgro™ L-glutamine Solution


Essential amino acid and a key component of culture media

Manufacturer: corning™  25005CV

 View more versions of this product

Catalog No. MT25005CV

  • / Pack of 6 Each
 

Description & Specifications

Specifications

pH 5.5 ±0.5
Format Liquid
Quantity 6 x 500mL
Sterilization Method Sterile-filtered
Packaging Type Plastic Bottle
Packaging Plastic Bottle
Solution Type L-Glutamine, 200mM solution (100X)
Storage Requirements 2° to 8°C
Quantity 6 x 500mL