Corning™ Improved MEM (Richter's Mod.), 1X
With L-Glutamine
Supplier:  Corning™ 10026CV
 View more versions of this product
Catalog No. MT10026CV
Description
Description
A modified formulation of Minimal Essential Medium, Improved Minimum Essential Medium (IMEM) contains a higher concentration of many components used in MEM along with additional amino acids, vitamins, and metabolites.
Specifications
Specifications
2° to 8°C
No Phenol Red
6 x 500 mL
MEM Richters
7 to 7.4