Corning™ Ham's Medium
Sterile, with L-Glutamine
Supplier: Corning™ 10080CM
Description
Lower documented Endotoxin content than offerings from other major suppliers. Ham's F-12 media was originally formulated for the serum-free culture of CHO cells, and is based on Ham's F-10 medium with increased concentrations of choline, inositol, putrescine, and several amino acids. This medium is also suitable for carcinoma cells, rat skeletal myoblasts, Chinese hamster lung cells, and rat, rabbit, and chicken embryos.Specifications
2° to 8°C | |
Sterile | |
7 to 7.4 |
Ham's F-12 Medium (Mod.) 1 X | |
Glutamine | |
6 x 1 L |