Corning™ G418 Sulfate, 50mg/mL
Blocks polypeptide synthesis by hindering protein elongation. Corning™ G418 Sulfate is an aminoglycoside similar in structure to gentamicin, neomycin and kanamycin.
Supplier: Corning™ 30234CI
Description
Well-known, effective selection agent G418 Sulfate is recommended for use as selection agent in a concentration range of 100-800 μg/mL. This antibiotic blocks polypeptide synthesis by hindering protein elongation, and inhibits the selection and maintenance of eukaryotic cells stably transfected with the neomycin resistance gene. G418 is an aminoglycoside similar in structure to gentamicin, neomycin and kanamycin.Specifications
2° to 8°C | |
Liquid | |
Sterile | |
100 mL |
50 mg/mL | |
Antibiotic | |
Plastic Bottle |