missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ cellgro™ DMEM/Ham's F-12 50/50 Mix, Powder

Used for culture of variety of cell types under low serum conditions. Corning™ cellgro™ DMEM/Ham's F-12 50/50 Mix, Powder ensures improved stability.

Manufacturer:  Corning™ 90091PB

 View more versions of this product

Catalog No. MT90091PB


 

Specifications

Description

Media, Cell Culture; Corning; cellgro; DMEM/Ham's F-12 50/50 Mix; w/o sodium bicarbonate and L-glutamine; Powder; Qty: 1 x 10L

  • Modification of Basal Medium Eagle (BME) containing four-fold concentrations of the amino acids and vitamins found in BME
  • Ham's F-12 media is based on Ham's F-10 medium with increased concentrations of choline, inositol, putrescine, and several amino acids
  • Formulation is without sodium bicarbonate, L-glutamine and phenol red
Description & Specifications

Specifications

1 x 10L
DMEM/Ham's F-12 50/50 Mix
2° to 8°C
Powder
1X
Sodium Bicarbonate and L-Glutamine