Please login to your online account to display your discounted pricing

Corning™ cellgro™ DMEM/Ham's F-12 50/50 Mix, Powder


Used for culture of variety of cell types under low serum conditions. Corning™ cellgro™ DMEM/Ham's F-12 50/50 Mix, Powder ensures improved stability.

Manufacturer: corning™  90090PB

 View more versions of this product

Catalog No. MT90090PB

  • / Each
 

Description & Specifications

Specifications

Product Type DMEM/Ham's F-12 50/50 Mix
Quantity 1 x 10L
Concentration 1X
Format Powder
Storage Requirements 2° to 8°C
Without Additives Sodium Bicarbonate, L-Glutamine, Phenol Red
Without Additives Sodium Bicarbonate, L-Glutamine, Phenol Red
Quantity 10000mL

  • Modification of Basal Medium Eagle (BME) containing four-fold concentrations of the amino acids and vitamins found in BME
  • Ham's F-12 media is based on Ham's F-10 medium with increased concentrations of choline, inositol, putrescine, and several amino acids
  • Formulation is without sodium bicarbonate, L-glutamine and phenol red